Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD11B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587247 Supplier Invitrogen™ Supplier No. PA587247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

HSD11B1 is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, HSD11B1 can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD11B1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Hsd11b1
Gene Accession No. P28845, P50172
Gene Alias 11 beta-hydroxysteroid dehydrogenase type 1; 11beta-HSD type 1; 11-beta-HSD1; 11beta-HSD1; 11beta-HSD-1; 11beta-HSD1A; 11-beta-hydroxysteroid dehydrogenase; 11-beta-hydroxysteroid dehydrogenase 1; 11-beta-hydroxysteroid dehydrogenase type 1; 11beta-hydroxysteroid dehydrogenase type 1; 11-DH; Corticosteroid 11-beta-dehydrogenase isozyme 1; Corticosteroid 11-beta-dehydrogenase, isozyme 1 (11-DH) (11-beta-hydroxysteroid dehydrogenase 1) (11-beta-HSD1) (11beta-HSD1A); cortocosteroid 11-beta dehydrogenase; CORTRD2; HDL; HSD11; HSD11B; Hsd11b1; HSD11L; hydroxysteroid (11-beta) dehydrogenase 1; hydroxysteroid 11-beta dehydrogenase 1; hydroxysteroid dehydrogenase, 11 beta type 1; LRRGT00065; MGC13539; OTTHUMP00000034650; SDR26C1; short chain dehydrogenase/reductase family 26C member 1; short chain dehydrogenase/reductase family 26C, member 1; short-chain reductase; short-chain reductase/dehydrogenase
Gene Symbols Hsd11b1
Host Species Rabbit
Immunogen Recombinant full length Human HSD11B1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15483, 3290
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.