Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD11B1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | HSD11B1L |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
HSD11B1L Polyclonal specifically detects HSD11B1L in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HSD11B1L | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
374875 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DYLVLNHIGGAPAGTRARSPQATRWLMQVNFVSYVQLTSRALPSLTDSKGSLVVVSSLLGRVPTSFSTPYS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
11-beta-hydroxysteroid dehydrogenase type 3, 11-DH3, EC 1.1.1, EC 1.1.1.-, HSD3,11-beta-HSD3, hydroxysteroid (11-beta) dehydrogenase 1-like, hydroxysteroid 11-beta-dehydrogenase 1-like protein, SCDR10short chain dehydrogenase/reductase 10, SDR26C2, short chain dehydrogenase/reductase family 26C, member 2, Short-chain dehydrogenase/reductase 10 | |
HSD11B1L | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title