Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD11B1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565543
Description
Immunogen sequence: DYLVLNHIGGA PAGTRARSPQ ATRWLMQVNF VSYVQLTSRA LPSLTDSKGS LVVVSSLLGR VPTSFSTPYS Highest antigen sequence identity to the following orthologs - mouse 41%, rat 38%.
This gene is a member of the hydroxysteroid dehydrogenase family. The encoded protein is similar to an enzyme that catalyzes the interconversion of inactive to active glucocorticoids (e.g. cortisone). Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
HSD11B1L | |
Polyclonal | |
Unconjugated | |
HSD11B1L | |
11-beta-HSD3; 11-beta-hydroxysteroid dehydrogenase type 3; 11-DH3; HSD11B1L; HSD3; hydroxysteroid (11-beta) dehydrogenase 1-like; hydroxysteroid 11-beta dehydrogenase 1 like; hydroxysteroid 11-beta-dehydrogenase 1-like protein; SCDR10; SCDR10B; SDR26C2; short chain dehydrogenase/reductase 10; short chain dehydrogenase/reductase family 26C member 2; short chain dehydrogenase/reductase family 26C, member 2; Short-chain dehydrogenase/reductase 10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374875 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5J1 | |
HSD11B1L | |
Recombinant Human HSD11B1L. Recombinant protein control fragment (Product #RP-106217). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction