Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD11B2 Antibody

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB10075626
Description
HSD11B2 Polyclonal specifically detects HSD11B2 in Human, Rat, Primate samples. It is validated for Western Blot.Specifications
HSD11B2 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
P80365 | |
HSD11B2 | |
The immunogen for this antibody was a sequence from 181a.a. to 405a.a. from HSD11B2(a 405a.a. long protein) | |
0.05 mg | |
Stem Cell Markers | |
3291 | |
Human, Rat, Primate |
Western Blot | |
Unconjugated | |
PBS with No Preservative | |
AME, corticosteroid 11-beta-dehydrogenase isozyme 2,11-beta-HSD2, EC 1.1.1, EC 1.1.1.-, HSD11KAME1, HSD2,11-DH2, hydroxysteroid (11-beta) dehydrogenase 2, NAD-dependent 11-beta-hydroxysteroid dehydrogenase, SDR9C3, short chain dehydrogenase/reductase family 9C member 3,11-beta-hydroxysteroid dehydrogenase type 2 | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
HSD11B2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction