Learn More
Invitrogen™ HSD11B2 Polyclonal Antibody

Description
HSD11B2 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | HSD11B2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | HSD11B2 |
| Gene Accession No. | P80365 |
| Gene Alias | 11(beta)-HSD2; 11-beta-HSD; 11-beta-HSD type II; 11-beta-HSD2; 11-beta-hydroxysteroid dehydrogenase type 2; 11-beta-hydroxysteroid dehydrogenase type II; 11-DH2; 11-HSD type II; 11HSD2; AME; AME1; corticosteroid 11-beta-dehydrogenase isozyme 2; -HSD11 type II; HSD11B2; HSD11K; HSD2; hydroxysteroid (11-beta) dehydrogenase 2; hydroxysteroid 11-beta dehydrogenase 2; Hydroxysteroid dehydrogenase, 11 beta type 2; NAD-dependent 11-beta-hydroxysteroid dehydrogenase; SDR9C3; Short chain dehydrogenase/reductase family 9C member 3 |
| Gene Symbols | HSD11B2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.