Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542058 Supplier Invitrogen™ Supplier No. PA542058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MARTVVLITG CSSGIGLHLA VRLASDPSQS FKVYATLRDL KTQGRLWEAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 91%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

HSD17B1 is favors the reduction of estrogens and androgens. It also has 20-alpha-HSD activity. It uses preferentially NADH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HSD17B1
Gene Accession No. P14061, P51656
Gene Alias 17-beta-dehydrogenase 1; 17beta-HSD; 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 17beta-hydroxysteroid dehydrogenase type 1; 17BHD1; 17HSDB1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E17KSR; E2DH; Edh17b1; EDH17B2; EDHB17; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; HSD17; HSD17B1; Hsd17ba; hydroxysteroid (17-beta) dehydrogenase 1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; hydroxysteroid 17-beta dehydrogenase 1; Hydroxysteroid dehydrogenase 17 beta, type 1; LOW QUALITY PROTEIN: estradiol 17-beta-dehydrogenase 1; placental 17-beta-hydroxysteroid dehydrogenase; SDR28C1; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1
Gene Symbols HSD17B1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HSD17B1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15485, 3292
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.