Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579400
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579400 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579400 Supplier Invitrogen™ Supplier No. PA579400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human T47D whole cell, human 293T whole cell, rat C6 whole cell. IHC: Rat Liver tissue, Rat Ovary tissue, Human Placenta tissue. ICC/IF: Hela cell.

HSD17B1 is favors the reduction of estrogens and androgens. It also has 20-alpha-HSD activity. It uses preferentially NADH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene HSD17B1
Gene Accession No. P14061, P51657
Gene Alias 17-beta-dehydrogenase 1; 17beta-HSD; 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 17beta-hydroxysteroid dehydrogenase type 1; 17BHD1; 17HSDB1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E17KSR; E2DH; Edh17b1; EDH17B2; EDHB17; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; HSD17; HSD17B1; Hsd17ba; hydroxysteroid (17-beta) dehydrogenase 1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; hydroxysteroid 17-beta dehydrogenase 1; Hydroxysteroid dehydrogenase 17 beta, type 1; LOW QUALITY PROTEIN: estradiol 17-beta-dehydrogenase 1; placental 17-beta-hydroxysteroid dehydrogenase; SDR28C1; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1
Gene Symbols HSD17B1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human HSD17B1 (29-43aa QSFKVYATLRDLKTQ).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25322, 3292
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.