Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B10 Mouse anti-Mouse, Human, Unconjugated, Clone: 5F3, MilliporeSigma™

Catalog No. MABN241625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2416-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2416-25 Supplier MilliporeSigma Supplier No. MABN241625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HSD17B10 antibody, clone 5F3, Cat. No. MABN2416, is a highly specific mouse monoclonal antibody that targets HSD17B10 and has been tested for use in Electron Microscopy, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B10
Applications Immunomicroscopy, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5F3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias 3-hydroxyacyl-CoA dehydrogenase type-2;17-beta-hydroxysteroid dehydrogenase 10;17-beta-HSD 10;3-hydroxy-2-methylbutyryl-CoA dehydrogenase;3-hydroxyacyl-CoA dehydrogenase type II;Endoplasmic reticulum-associated amyloid beta-peptide-binding protein;Mitochondrial ribonuclease P protein 2;Mitochondrial RNase P protein 2;Short-chain type dehydrogenase/reductase XH98G2;Type II HADH
Gene Symbols HSD17B10;ERAB;HADH2;MRPP2;SCHAD;SDR5C1;XH98G2
Host Species Mouse
Immunogen GST-tagged full length recombinant human 3-hydroxyacyl-CoA dehydrogenase type-2 protein (ERAB/HADH).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 5F3 detects 3-hydroxyacyl-CoA dehydrogenase type-2 in human and murine cells.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.