Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD17B11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554469
Description
Immunogen sequence: FVNTGFIKNP STSLGPTLEP EEVVNRLMHG ILTEQKMIFI PSSIAFLTTL ERILPERFLA VLKRKISVKF DAVIGY Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.
Specifications
| HSD17B11 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B11 | |
| 17-beta-HSD 11; 17-beta-HSD XI; 17betaHSD11; 17-BETA-HSD11; 17beta-HSD11; 17betaHSDXI; 17-BETA-HSDXI; 17-beta-hydroxysteroid dehydrogenase 11; 17-beta-hydroxysteroid dehydrogenase type XI; 17-beta-hydroxysteroid dehydrogenase XI; 17bHSD11; CTCL tumor antigen HD-CL-03; CTCL-associated antigen HD-CL-03; Cutaneous T-cell lymphoma-associated antigen HD-CL-03; dehydrogenase/reductase (SDR family) member 8; dehydrogenase/reductase SDR family member 8; Dhrs8; estradiol 17-beta-dehydrogenase 11; estradiol 17-beta-dehydrogenase 11-like protein; Hsd17b11; hydroxysteroid (17-beta) dehydrogenase 11; hydroxysteroid 17-beta dehydrogenase 11; MGC137132 protein; PAN1B; PSEC0029; Retinal short-chain dehydrogenase/reductase 2; retinal short-chain dehydrogenase/reductase SDR2; retSDR2; SDR16C2; SDR2; short chain dehydrogenase/reductase family 16C member 2; short chain dehydrogenase/reductase family 16C, member 2; UNQ207/PRO233 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51170 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NBQ5 | |
| HSD17B11 | |
| Recombinant protein corresponding to Human HSD17B11. Recombinant protein control fragment (Product #RP-93159). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction