Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B12 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550424

Catalog No. PA550424


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total HSD17B12 protein.

The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSD17B12
Polyclonal
Unconjugated
HSD17B12
17-beta-HSD 12; 17beta-HSD type 12; 17-beta-hydroxysteroid dehydrogenase 12; 2610510O05Rik; 3-ketoacyl-CoA reductase; AI172963; Estradiol 17-beta-dehydrogenase 12; HSD17B12; hydroxysteroid (17-beta) dehydrogenase 12; hydroxysteroid 17-beta dehydrogenase 12; KAR; keratoadhesin; keratonectin; Kik1; KIK-I; SDR12C1; short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3; steroid dehydrogenase; steroid dehydrogenase homolog; very-long-chain 3-oxoacyl-CoA reductase
Rabbit
Antigen affinity chromatography
RUO
51144
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q53GQ0
HSD17B12
Fusion protein corresponding to a region derived from internal residues of human hydroxysteroid (17-beta) dehydrogenase 12.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.