Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595888 Supplier Invitrogen™ Supplier No. PA595888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGVGPGLGEW AVVTGSTDGI GKSYAEELAK HGMKVVLISR SKDKLDQVSS EIKEKFKVET RTIAVDFASE DIYDKIKTGL AGLEIGILVN NVGMSYEYPE YFLDVPDLDN VIKKMININI LSVCKMTQLV LPGMVERSKG AILNISSGSG MLPVPLLTIY SATKTFVDFF SQCLHEEYRS KGVFVQSVLP YFVATKLAKI RKPTLDKPSP ETFVKSAIKT VGLQSRTNGY LIHALMGSII SNLPSWIYLK IVMNMNKSTR AHYLKKTKKN.

The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HSD17B12
Gene Accession No. Q53GQ0
Gene Alias 17-beta-HSD 12; 17beta-HSD type 12; 17-beta-hydroxysteroid dehydrogenase 12; 2610510O05Rik; 3-ketoacyl-CoA reductase; AI172963; Estradiol 17-beta-dehydrogenase 12; HSD17B12; hydroxysteroid (17-beta) dehydrogenase 12; hydroxysteroid 17-beta dehydrogenase 12; KAR; keratoadhesin; keratonectin; Kik1; KIK-I; SDR12C1; short chain dehydrogenase/reductase family 12C member 1; short chain dehydrogenase/reductase family 12C, member 1; smooth muscle-specific 17 beta-hydroxysteroid dehydrogenase type 3; steroid dehydrogenase; steroid dehydrogenase homolog; very-long-chain 3-oxoacyl-CoA reductase
Gene Symbols HSD17B12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 43-312 of human HSD17B12 (NP_0572261).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51144
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.