Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD17B13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556063
Description
Immunogen sequence: NNAGTVYPAD LLSTKDEEIT KTFEVNILGH FWITKALLPS MMERNHGHIV TVASVCGHEG IPYLIPYCSS KFAAVGFHRG LTSELQALGK TGIKTSCLCP VFVNTGFTKN PSTRLWPVLE TDEVVRSLI Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
HSD17B13 (Hydroxysteroid 17 Beta Dehydrogenase 13) is a Protein Coding gene. Diseases associated with HSD17B13 include5 Alpha Reductase DeficiencyandMucopolysaccharidosis Iii. Among its related pathways areMetabolismandLipoprotein metabolism. Gene Ontology (GO) annotations related to this gene includeoxidoreductase activity. An important paralog of this gene isHSD17B11.
Specifications
HSD17B13 | |
Polyclonal | |
Unconjugated | |
HSD17B13 | |
17-beta hydroxysteroid dehydrogenase; 17-beta hydroxysteroid dehydrogenase 13; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; AI047820; Alcohol dehydrogenase PAN1B-like; HMFN0376; Hsd17b13; hydroxysteroid (17-beta) dehydrogenase 13; hydroxysteroid 17-beta dehydrogenase 13; NIIL497; Pan1b; PAN1B-like; SCDR9; SDR16C3; short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; short-chain dehydrogenase/reductase 9; UNQ497/PRO1014 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
345275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5P4 | |
HSD17B13 | |
Recombinant protein corresponding to Human HSD17B13. Recombinant protein control fragment (Product #RP-90460). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction