Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B13 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA525633

Catalog No. PIPA525633


Only null left
Add to Cart

Description

Description

HSD17B13 Polyclonal Antibody for Western Blot, IHC (P)

HSD17B13 (Hydroxysteroid 17 Beta Dehydrogenase 13) is a Protein Coding gene. Diseases associated with HSD17B13 include5 Alpha Reductase DeficiencyandMucopolysaccharidosis Iii. Among its related pathways areMetabolismandLipoprotein metabolism. Gene Ontology (GO) annotations related to this gene includeoxidoreductase activity. An important paralog of this gene isHSD17B11.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSD17B13
Polyclonal
Unconjugated
HSD17B13
17-beta hydroxysteroid dehydrogenase; 17-beta hydroxysteroid dehydrogenase 13; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; AI047820; Alcohol dehydrogenase PAN1B-like; HMFN0376; Hsd17b13; hydroxysteroid (17-beta) dehydrogenase 13; hydroxysteroid 17-beta dehydrogenase 13; NIIL497; Pan1b; PAN1B-like; SCDR9; SDR16C3; short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; short-chain dehydrogenase/reductase 9; UNQ497/PRO1014
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
345275
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.7 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q7Z5P4
HSD17B13
KLH conjugated synthetic peptide between 81-110 amino acids from the N-terminal region of human DHB13.
200 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.