Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544634 Supplier Thermo Scientific Supplier No. PA544634
Add to Cart
Add to Cart

HSD17B14 Polyclonal Antibody

HSD17B14 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HSD17B14
Gene Accession No. Q9BPX1
Gene Alias 0610039E24Rik; 17-beta-HSD 14; 17-beta-hydroxysteroid dehydrogenase 14; 17-beta-hydroxysteroid dehydrogenase DHRS10; dehydrogenase/reductase (SDR family) member 10; dehydrogenase/reductase SDR family member 10; DHRS10; HSD17B14; hydroxysteroid (17-beta) dehydrogenase 14; hydroxysteroid 17-beta dehydrogenase 14; retinal short-chain dehydrogenase/reductase 3; retinal short-chain dehydrogenase/reductase retSDR3; retSDR3; SDR3; SDR47C1; Short chain dehydrogenase/reductase family 47C member 1; short chain dehydrogenase/reductase family 47C, member 1; testicular tissue protein Li 52; UNQ502/PRO474
Gene Symbols HSD17B14
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HSD17B14.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51171
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.