Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554417 Supplier Invitrogen™ Supplier No. PA554417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATGTRYAGKV VVVTGGGRGI GAGIVRAFVN SGARVVICDK DESGGRALEQ ELPGAVFILC DVTQEDDVKT LVSETIR Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 71%.

17-beta-hydroxysteroid dehydrogenases, such as HSD17B14, are primarily involved in metabolism of steroids at the C17 position and also of other substrates, such as fatty acids, prostaglandins, and xenobiotics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSD17B14
Gene Accession No. Q9BPX1
Gene Alias 0610039E24Rik; 17-beta-HSD 14; 17-beta-hydroxysteroid dehydrogenase 14; 17-beta-hydroxysteroid dehydrogenase DHRS10; dehydrogenase/reductase (SDR family) member 10; dehydrogenase/reductase SDR family member 10; DHRS10; HSD17B14; hydroxysteroid (17-beta) dehydrogenase 14; hydroxysteroid 17-beta dehydrogenase 14; retinal short-chain dehydrogenase/reductase 3; retinal short-chain dehydrogenase/reductase retSDR3; retSDR3; SDR3; SDR47C1; Short chain dehydrogenase/reductase family 47C member 1; short chain dehydrogenase/reductase family 47C, member 1; testicular tissue protein Li 52; UNQ502/PRO474
Gene Symbols HSD17B14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HSD17B14. Recombinant protein control fragment (Product #RP-93124).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51171
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.