Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD17B2 Antibody (OTI4F10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270945
Description
HSD17B2 Monoclonal specifically detects HSD17B2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
HSD17B2 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Flow Cytometry 1:100, CyTOF-ready | |
17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, E2DH, EC 1.1.1.62,17-beta-HSD 2, EC 1.1.1.63, EDH17B220-alpha-HSD, estradiol 17-beta-dehydrogenase 2, HSD17, hydroxysteroid (17-beta) dehydrogenase 2, Microsomal 17-beta-hydroxysteroid dehydrogenase, SDR9C2, short chain dehydrogenase/reductase family 9C, member 2, Testosterone 17-beta-dehydrogenase | |
Mouse | |
42.6 kDa | |
100 μg | |
3294 | |
Human | |
Lyophilized |
Western Blot, Flow Cytometry, CyTOF | |
OTI4F10 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
HSD17B2 | |
Full length human recombinant protein of human HSD17B2(NP_002144) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction