Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD17B2 Antibody (OTI4F10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270945H
Description
HSD17B2 Monoclonal specifically detects HSD17B2 in Human samples. It is validated for Western Blot.Specifications
HSD17B2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HSD17B2 | |
Full length recombinant protein of human HSD17B2 (NP_002144) produced in HEK293T cells. | |
0.1 mL | |
3294.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4F10 | |
Western Blot | |
17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, E2DH, EC 1.1.1.62,17-beta-HSD 2, EC 1.1.1.63, EDH17B220-alpha-HSD, estradiol 17-beta-dehydrogenase 2, HSD17, hydroxysteroid (17-beta) dehydrogenase 2, Microsomal 17-beta-hydroxysteroid dehydrogenase, SDR9C2, short chain dehydrogenase/reductase family 9C, member 2, Testosterone 17-beta-dehydrogenase | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction