Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576648 Supplier Invitrogen™ Supplier No. PA576648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B2
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene HSD17B2
Gene Accession No. P37059, P51658, Q62730
Gene Alias 1.1.1.239; 17 beta HSD 2; 17 HSD type 2; 17-beta hydroxysteroid dehydrogenase type 2; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha HSD; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; AI194836; AI194967; AI255511; E2DH; EDH17B2; estradiol 17-beta-dehydrogenase 2; HSD17; HSD17B2; hydroxysteroid (17-beta) dehydrogenase 2; hydroxysteroid 17-beta dehydrogenase 2; microsomal 17-beta-hydroxysteroid dehydrogenase; SDR9C2; short chain dehydrogenase/reductase family 9C member 2; short chain dehydrogenase/reductase family 9C, member 2; tes; testosterone 17-beta-dehydrogenase
Gene Symbols HSD17B2
Host Species Rabbit
Immunogen Recombinant full length Human HSD17B2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15486, 3294, 79243
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.