Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD17B2 Monoclonal Antibody (OTI3E9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525937
Description
HSD17B2 Monoclonal Antibody for Western Blot, Flow
HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.
Specifications
HSD17B2 | |
Monoclonal | |
0.88 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P37059 | |
HSD17B2 | |
Full length human recombinant protein of HSD17B2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI3E9 | |
Unconjugated | |
HSD17B2 | |
1.1.1.239; 17 beta HSD 2; 17 HSD type 2; 17-beta hydroxysteroid dehydrogenase type 2; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha HSD; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; AI194836; AI194967; AI255511; E2DH; EDH17B2; estradiol 17-beta-dehydrogenase 2; HSD17; HSD17B2; hydroxysteroid (17-beta) dehydrogenase 2; hydroxysteroid 17-beta dehydrogenase 2; microsomal 17-beta-hydroxysteroid dehydrogenase; SDR9C2; short chain dehydrogenase/reductase family 9C member 2; short chain dehydrogenase/reductase family 9C, member 2; tes; testosterone 17-beta-dehydrogenase | |
Mouse | |
Affinity chromatography | |
RUO | |
3294 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction