Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563356 Supplier Invitrogen™ Supplier No. PA563356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVGILPNLLP SHFLNAPDEI QSLIHCNITS VVKMTQLILK HMESRQKGLI L Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predominantly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSD17B3
Gene Accession No. P37058
Gene Alias 17(beta)HSD type 3; 17beta-HSD; 17-beta-HSD 3; 17-beta-HSD3; 17-beta-hydroxysteroid dehydrogenase type 3; Edh17b3; estradiol 17 beta-dehydrogenase 3; HSD17B3; hydroxysteroid (17-beta) dehydrogenase 3; hydroxysteroid 17-beta dehydrogenase 3; hydroxysteroid dehydrogenase 3; SDR12C2; Short chain dehydrogenase/reductase family 12C member 2; short chain dehydrogenase/reductase family 12C, member 2; testicular 17-beta-hydroxysteroid dehydrogenase; Testosterone 17-beta-dehydrogenase 3
Gene Symbols HSD17B3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HSD17B3. Recombinant protein control fragment (Product #RP-103273).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3293
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.