Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD17B3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143264
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predominantly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia.
Specifications
| HSD17B3 | |
| Polyclonal | |
| Unconjugated | |
| HSD17B3 | |
| 17(beta)HSD type 3; 17beta-HSD; 17-beta-HSD 3; 17-beta-HSD3; 17-beta-hydroxysteroid dehydrogenase type 3; Edh17b3; estradiol 17 beta-dehydrogenase 3; HSD17B3; hydroxysteroid (17-beta) dehydrogenase 3; hydroxysteroid 17-beta dehydrogenase 3; hydroxysteroid dehydrogenase 3; SDR12C2; Short chain dehydrogenase/reductase family 12C member 2; short chain dehydrogenase/reductase family 12C, member 2; testicular 17-beta-hydroxysteroid dehydrogenase; Testosterone 17-beta-dehydrogenase 3 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 3293 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P37058 | |
| HSD17B3 | |
| Peptide with sequence C-SHFLNAPDEIQSLIH (aa 141-155). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction