Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143917
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143917 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143917 Supplier Invitrogen™ Supplier No. PA5143917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human U20S whole cell, human HEL whole cell, rat testis tissue, rat liver tissue, mouse liver tissue, mouse RAW2647 whole cell. Flow: HepG2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predominantly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene HSD17B3
Gene Accession No. O54939, P37058, P70385
Gene Alias 17(beta)HSD type 3; 17beta-HSD; 17-beta-HSD 3; 17-beta-HSD3; 17-beta-hydroxysteroid dehydrogenase type 3; Edh17b3; estradiol 17 beta-dehydrogenase 3; HSD17B3; hydroxysteroid (17-beta) dehydrogenase 3; hydroxysteroid 17-beta dehydrogenase 3; hydroxysteroid dehydrogenase 3; SDR12C2; Short chain dehydrogenase/reductase family 12C member 2; short chain dehydrogenase/reductase family 12C, member 2; testicular 17-beta-hydroxysteroid dehydrogenase; Testosterone 17-beta-dehydrogenase 3
Gene Symbols HSD17B3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human HSD17B3 (NLLPSHFLNAPDEIQSLIH).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117182, 15487, 3293
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.