Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B4/MFE-2/17-beta-HSD4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568703
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8703 Supplier Bethyl Laboratories, Inc Supplier No. A304797A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Peroxisomal multifunctional enzyme type 2 is a protein that in humans is encoded by the HSD17B4 gene. The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen HSD17B4/MFE-2/17-beta-HSD4
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene HSD17B4
Gene Accession No. P51659
Gene Alias DBP, MFE-2, MPF-2, SDR8C1, 17-beta-HSD 4, 17-beta-HSD IV, 17-beta-hydroxysteroid dehydrogenase 4, 17beta-estradiol dehydrogenase type IV, 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase, D-3-hydroxyacyl-CoA dehydratase, D-bifunctional protein, peroxisomal, beta-hydroxyacyl dehydrogenase, beta-keto-reductase, multifunctional protein 2, peroxisomal multifunctional enzyme type 2, peroxisomal multifunctional protein 2, short chain dehydrogenase/reductase family 8C, member 1
Gene Symbols HSD17B4
Host Species Rabbit
Immunogen Between 600 and 650
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3295
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.