Learn More
HSD17B4/MFE-2/17-beta-HSD4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304797AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Peroxisomal multifunctional enzyme type 2 is a protein that in humans is encoded by the HSD17B4 gene. The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
HSD17B4/MFE-2/17-beta-HSD4 | |
Polyclonal | |
Unconjugated | |
HSD17B4 | |
DBP, MFE-2, MPF-2, SDR8C1, 17-beta-HSD 4, 17-beta-HSD IV, 17-beta-hydroxysteroid dehydrogenase 4, 17beta-estradiol dehydrogenase type IV, 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase, D-3-hydroxyacyl-CoA dehydratase, D-bifunctional protein, peroxisomal, beta-hydroxyacyl dehydrogenase, beta-keto-reductase, multifunctional protein 2, peroxisomal multifunctional enzyme type 2, peroxisomal multifunctional protein 2, short chain dehydrogenase/reductase family 8C, member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3295 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P51659 | |
HSD17B4 | |
Between 600 and 650 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.