Learn More
Invitrogen™ HSD17B4 Monoclonal Antibody (OTI1F3)

Description
HSD17B4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | HSD17B4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1F3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HSD17B4 |
| Gene Accession No. | P51659 |
| Gene Alias | (3R)-hydroxyacyl-CoA dehydrogenase; 17[b]-HSD; 17beta-estradiol dehydrogenase type IV; 17-beta-HSD; 17-beta-HSD 4; 17-beta-HSD IV; 17-beta-hydroxysteroid dehydrogenase 4; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase; 4; beta-hydroxyacyl dehydrogenase; beta-keto-reductase; D-3-hydroxyacyl-CoA dehydratase; D-bifunctional protein; D-bifunctional protein, peroxisomal; DBP; Edh17b4; Enoyl-CoA hydratase 2; Hsd17b4; hydroxysteroid (17-beta) dehydrogenase 4; hydroxysteroid 17-beta dehydrogenase 4; hydroxysteroid dehydrogenase 4; MFE-2; MFP2; Mfp-2; MPF-2; Multifunctional protein 2; perMFE-2; peroxisomal multifunctional enzyme type 2; peroxisomal multifunctional enzyme type II; peroxisomal multifunctional protein 2; PRLTS1; SDR8C1; Short chain dehydrogenase/reductase family 8C member 1; short chain dehydrogenase/reductase family 8C, member 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.