Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B4 Mouse anti-Human, Clone: 8E5AB11, Abnova™

Catalog No. 89035537 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-537 Supplier Abnova Corporation Supplier No. MAB14139

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human HSD17B4.

  • High specificity
  • High reactivity

Specifications

Antigen HSD17B4
Applications ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 8E5AB11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human HSD17B4.
Dilution ELISA Immunocytochemistry (4 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. P51659
Gene Alias DBP/MFE-2/SDR8C1
Gene Symbols HSD17B4
Host Species Mouse
Immunogen Human HSD17B4.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3295
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.