Learn More
HSD17B4 Mouse anti-Human, Clone: OTI4F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF507027

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Peroxisomal multifunctional enzyme type 2 is a protein that in humans is encoded by the HSD17B4 gene. The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
HSD17B4 | |
Monoclonal | |
Unconjugated | |
HSD17B4 | |
DBP, MFE-2, MPF-2, PRLTS1, SDR8C1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3295 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P51659 | |
HSD17B4 | |
Full length human recombit protein of human HSD17B4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.