Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110873 Supplier Invitrogen™ Supplier No. PA5110873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNNAGILRDR SFARISDEDW DIIHRVHLRG SFQVTRAAWE HMKKQKYGRI IMTSSASGIY GNFGQANYSA AKLGLLGLAN SLAIEGRKSN IHCNTIAPNA GSRMTQTVMP EDLVEALKPE YVAPLV Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 93%.

Peroxisomal multifunctional enzyme type 2 is a protein that in humans is encoded by the HSD17B4 gene. The protein encoded by this gene is a bifunctional enzyme that is involved in the peroxisomal beta-oxidation pathway for fatty acids. It also acts as a catalyst for the formation of 3-ketoacyl-CoA intermediates from both straight-chain and 2-methyl-branched-chain fatty acids. Defects in this gene that affect the peroxisomal fatty acid beta-oxidation activity are a cause of D-bifunctional protein deficiency (DBPD). An apparent pseudogene of this gene is present on chromosome 8. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSD17B4
Gene Accession No. P51659
Gene Alias (3R)-hydroxyacyl-CoA dehydrogenase; 17[b]-HSD; 17beta-estradiol dehydrogenase type IV; 17-beta-HSD; 17-beta-HSD 4; 17-beta-HSD IV; 17-beta-hydroxysteroid dehydrogenase 4; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase; 4; beta-hydroxyacyl dehydrogenase; beta-keto-reductase; D-3-hydroxyacyl-CoA dehydratase; D-bifunctional protein; D-bifunctional protein, peroxisomal; DBP; Edh17b4; Enoyl-CoA hydratase 2; Hsd17b4; hydroxysteroid (17-beta) dehydrogenase 4; hydroxysteroid 17-beta dehydrogenase 4; hydroxysteroid dehydrogenase 4; MFE-2; MFP2; Mfp-2; MPF-2; Multifunctional protein 2; perMFE-2; peroxisomal multifunctional enzyme type 2; peroxisomal multifunctional enzyme type II; peroxisomal multifunctional protein 2; PRLTS1; SDR8C1; Short chain dehydrogenase/reductase family 8C member 1; short chain dehydrogenase/reductase family 8C, member 1
Gene Symbols HSD17B4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human HSD17B4. Recombinant protein control fragment (Product #RP-93145).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3295
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.