Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD17B6 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297586AF488
Description
HSD17B6 Polyclonal specifically detects HSD17B6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HSD17B6 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
17-beta-HSD 6, 17-beta-HSD6, 17-beta-hydroxysteroid dehydrogenase type 6, 3(alpha->beta)-hydroxysteroid epimerase, 3-alpha->beta-HSE, EC 1.1.1, EC 1.1.1.105, EC 1.1.1.62, EC 1.1.1.63, HSE3-hydroxysteroid epimerase, hydroxysteroid (17-beta) dehydrogenase 6, hydroxysteroid (17-beta) dehydrogenase 6 homolog (mouse), NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase, Oxidative 3-alpha hydroxysteroid dehydrogenase, oxidative 3-alpha-hydroxysteroid-dehydrogenase, oxidoreductase, retinol dehydrogenase, RODH3(alpha->beta)-hydroxysteroid epimerasel, SDR9C6, short chain dehydrogenase/reductase family 9C, member 6,3-alpha->beta-hydroxysteroid epimerase | |
Produced in rabbits immunized with E. coli-derived Human HSD17B6 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8630 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction