Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD17B6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542018
Description
Peptide sequence: MWLYLAAFVG LYYLLHWYRE RQVVSHLQDK YVFITGCDSG FGNLLARQLD Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%.
The protein encoded by this gene has both oxidoreductase and epimerase activities and is involved in androgen catabolism. The oxidoreductase activity can convert 3 alpha-adiol to dihydrotestosterone, while the epimerase activity can convert androsterone to epi-androsterone. Both reactions use NAD+ as the preferred cofactor. This gene is a member of the retinol dehydrogenase family. Transcript variants utilizing alternative polyadenylation signals exist.
Specifications
HSD17B6 | |
Polyclonal | |
Unconjugated | |
HSD17B6 | |
17-beta-HSD 6; 17-beta-HSD6; 17betaHSD9; 17-beta-HSD9; 17-beta-hydroxysteroid dehydrogenase type 6; 3(alpha->beta)-hydroxysteroid epimerase; 3(alpha->beta)-hydroxysteroid epimerasel; 3-alpha->beta-HSE; 3-alpha->beta-hydroxysteroid epimerase; 3-hydroxysteroid epimerase; Gm182; Hsd17b6; Hsd17b9; HSE; hydroxysteroid (17-beta) dehydrogenase 6; hydroxysteroid (17-beta) dehydrogenase 6 homolog; hydroxysteroid (17-beta) dehydrogenase 9; hydroxysteroid 17-beta dehydrogenase 6; hydroxysteroid 17-beta dehydrogenase 9; hypothetical protein LOC533086; NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase; oxidative 17 beta hydroxysteroid dehydrogenase type 6; oxidative 3-alpha hydroxysteroid dehydrogenase; oxidative 3-alpha-hydroxysteroid-dehydrogenase; oxidoreductase; Rdh8; retinol dehydrogenase; RODH; SDR9C6; Short chain dehydrogenase/reductase family 9C member 6; short chain dehydrogenase/reductase family 9C, member 6 | |
Rabbit | |
Protein A | |
RUO | |
8630 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14756 | |
HSD17B6 | |
Synthetic peptide directed towards the N-terminal of human HSD17B6 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction