Learn More
Invitrogen™ HSD17B6 Polyclonal Antibody

Description
Immunogen sequence: VEPGYFRTGM TNMTQSLERM KQSWKEAPKH IKETYGQQYF DALYNIMKEG LL Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 44%.
Specifications
Specifications
| Antigen | HSD17B6 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HSD17B6 |
| Gene Accession No. | O14756 |
| Gene Alias | 17-beta-HSD 6; 17-beta-HSD6; 17betaHSD9; 17-beta-HSD9; 17-beta-hydroxysteroid dehydrogenase type 6; 3(alpha->beta)-hydroxysteroid epimerase; 3(alpha->beta)-hydroxysteroid epimerasel; 3-alpha->beta-HSE; 3-alpha->beta-hydroxysteroid epimerase; 3-hydroxysteroid epimerase; Gm182; Hsd17b6; Hsd17b9; HSE; hydroxysteroid (17-beta) dehydrogenase 6; hydroxysteroid (17-beta) dehydrogenase 6 homolog; hydroxysteroid (17-beta) dehydrogenase 9; hydroxysteroid 17-beta dehydrogenase 6; hydroxysteroid 17-beta dehydrogenase 9; hypothetical protein LOC533086; NAD+ -dependent 3 alpha-hydroxysteroid dehydrogenase 3-hydroxysteroid epimerase; oxidative 17 beta hydroxysteroid dehydrogenase type 6; oxidative 3-alpha hydroxysteroid dehydrogenase; oxidative 3-alpha-hydroxysteroid-dehydrogenase; oxidoreductase; Rdh8; retinol dehydrogenase; RODH; SDR9C6; Short chain dehydrogenase/reductase family 9C member 6; short chain dehydrogenase/reductase family 9C, member 6 |
| Gene Symbols | HSD17B6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.