Learn More
Invitrogen™ HSD17B7 Polyclonal Antibody

Description
Positive test controls include: Mouse liver, Mouse ovary. The target is usually found in the following locations: Cell membrane, Single-pass membrane protein. Immunogen sequence: SRKVIHMFST AEGLLTQGDK ITADGLQEVF ETNVFGHFIL IRELEPLLCH SDNPSQLIWT SSRSARKSNF SLEDFQHSKG KEPYSSSKYA T.
Specifications
Specifications
| Antigen | HSD17B7 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | HSD17B7 |
| Gene Accession No. | O88736, P56937 |
| Gene Alias | 17 beta-hydroxysteroid dehydrogenase type VII; 17beta hydroxysteroid dehydrogenase; 17-beta-HSD 7; 17-beta-hydroxysteroid dehydrogenase 7; 3-keto-steroid reductase; AI266814; bA291L22.1; ERG27; Estradiol 17-beta-dehydrogenase 7; Hsd17b_2; HSD17B7; HSD17B7P2; hydroxysteroid (17-beta) dehydrogenase 7; hydroxysteroid 17-beta dehydrogenase 7; hydroxysteroid 17-beta dehydrogenase 7 pseudogene 2; hydroxysteroid dehydrogenase 17 beta, type 7; PRAP; PRL receptor-associated protein; SDR37C1; Short chain dehydrogenase/reductase family 37C member 1; short chain dehydrogenase/reductase family 37C, member 1; UNQ2563/PRO6243 |
| Gene Symbols | HSD17B7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.