Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD17B8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550423
Description
The antibody detects endogenous levels of total HSD17B8 protein.
NAD-dependent 17-beta-hydroxysteroid dehydrogenase with highest activity towards estradiol. Has very low activity towards testosterone. The heteroteramer with CBR4 has NADH-dependent 3- ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria.
Specifications
HSD17B8 | |
Polyclonal | |
Unconjugated | |
HSD17B8 | |
17-beta-HSD 8; 17-beta-hydroxysteroid dehydrogenase 8; 3-ketoacyl-[acyl-carrier-protein] reductase alpha subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; D17H6S112E; D6S2245E; dJ1033B10.9; Estradiol 17-beta-dehydrogenase 8; estrogen 17-oxidoreductase; FABG; FABGL; H FABGL; H2-K region expressed gene 6; H-2Ke6; H2-KE6; Hke6; Hsd17b8; hydroxysteroid (17-beta) dehydrogenase 8; hydroxysteroid 17-beta dehydrogenase 8; KAR alpha subunit; KE6; Ke-6; Protein Ke6; really interesting new gene 2 protein; RING2; SDR30C1; Short chain dehydrogenase/reductase family 30C member 1; short chain dehydrogenase/reductase family 30C, member 1; Testosterone 17-beta-dehydrogenase 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7923 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q92506 | |
HSD17B8 | |
Full length fusion protein of Human HSD17B8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction