Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD17B8 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11407R

Description
NAD-dependent 17-beta-hydroxysteroid dehydrogenase with highest activity towards estradiol. Has very low activity towards testosterone. The heteroteramer with CBR4 has NADH-dependent 3- ketoacyl-acyl carrier protein reductase activity. May play a role in biosynthesis of fatty acids in mitochondria.Specifications
HSD17B8 | |
Polyclonal | |
Unconjugated | |
HSD17B8 | |
17-beta-HSD 8; 17-beta-hydroxysteroid dehydrogenase 8; 3-ketoacyl-[acyl-carrier-protein] reductase alpha subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; D6S2245E; dJ1033B10.9; Estradiol 17-beta-dehydrogenase 8; estrogen 17-oxidoreductase; FABG; FABGL; H FABGL; H2-KE6; HKE6; HSD17B8; hydroxysteroid (17-beta) dehydrogenase 8; hydroxysteroid 17-beta dehydrogenase 8; KAR alpha subunit; Ke6; Ke-6; Protein Ke6; Really interesting new gene 2 protein; RING2; SDR30C1; Short chain dehydrogenase/reductase family 30C member 1; short chain dehydrogenase/reductase family 30C, member 1; Testosterone 17-beta-dehydrogenase 8 | |
Rabbit | |
Protein A | |
RUO | |
7923 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q92506 | |
HSD17B8 | |
KLH conjugated synthetic peptide derived from human HSD17B8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction