Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD3B1 Antibody (FDO66Q), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | HSD3B1 |
---|---|
Clone | FDO66Q |
Concentration | 1 mg/mL |
Dilution | Western Blot, Immunohistochemistry 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 10-20 ug/ml, Immunohistochemistry-Frozen 10-20 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11078644T
![]() |
Novus Biologicals
NB11078644SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11078644
![]() |
Novus Biologicals
NB11078644 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
HSD3B1 Monoclonal specifically detects HSD3B1 in Human, Mouse, Rat, Porcine, Bovine, Goat, Baboon, Primate, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
HSD3B1 | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Lipid and Metabolism, Prostate Cancer, Signal Transduction | |
3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3BETAHSD, 3-beta-HSD I, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase, 3BH, delta-5-3-ketosteroid isomerase, EC 1.1.1, HSD3B, HSDB3, HSDB3A, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1, hydroxy-delta-5-steroid dehydrogenase, 3 beta-and steroid delta-isomerase 1, I, progesterone reductase, SDR11E1, short chain dehydrogenase/reductase family 11E, member 1,3-beta-hydroxy-5-ene steroid dehydrogenase, steroid Delta-isomerase, Trophoblast antigen FDO161G | |
HSD3B1 | |
IgG1 | |
Protein G purified | |
Human, baboon, bovine, rat and marmoset. |
FDO66Q | |
Western Blot, Immunohistochemistry 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 10-20 ug/ml, Immunohistochemistry-Frozen 10-20 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Bovine, Goat, Baboon, Primate, Sheep | |
3283 | |
JEG Choriocarcinoma cells | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title