Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD3B1, Mouse anti-Human, Clone: 4C1, Millipore Sigma™

Catalog No. MABS206825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS206825 25 μg
MABS2068100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS206825 Supplier MilliporeSigma Supplier No. MABS206825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HSD3B1, clone 4C1, Cat. No. MABS2068, is a mouse monoclonal antibody that detects 3-beta-HSD I (HSD3B1) and has been tested for use in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.

3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 (UniProt: P14060; also known as 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3-beta-HSD I, Trophoblast antigen FDO161G) is encoded by the HSD3B1 (also known as 3BH, HSDB3A) gene (Gene ID: 3283) in human. HSD3B1 is a single -pass membrane protein localized in mitochondria and endoplasmic reticulum. Its transmembrane region is localized to amino acids 288-308. It is a bifunctional enzyme that catalyzes the oxidative conversion of Delta 5-ene-3-beta-hydroxy steroid and causes oxidative conversion of ketosteroids. It plays a vital role in the biosynthesis of all classes of hormonal steroids. Humans have two HSD3B isozymes. HSD3B1 is mainly expressed in the placenta, mammary and, prostate glands, and other peripheral tissues including the skin and adipose tissue and HSD3B2 is expressed primarily in the adrenal and gonads. They exhibit about 93% sequence homology. (Ref.: Gomez-Sanchez, CE et al. (2017). Steroids 127; 56-61).

TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD3B1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4C1
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected HSD3B1 in human adrenal gland tissue sections.ELISA Analysis : A representative lot detected HSD3B1 in ELISA applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).Western Blotting Analysis: 1μg/mL from a representative lot detected HSD3B1 in human placenta tissue lysate in Western blotting applications.Immunohistochemistry (Paraffin) Analysis: A representative lot detected HSD3B1 in Immunohistochemistry applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).Western Blotting Analysis: A representative lot detected HSD3B1 in Western Blotting applications (Gomez-Sanchez, C.E., et. al. (2017). Steroids. 127:56-61).
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1;3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I;3-beta-HSD I;Trophoblast antigen FDO161G
Gene Symbols HSD3B1;3BH;HSD3BA
Host Species Mouse
Immunogen A KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of human 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 (HSD3B1).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000853.1
Test Specificity Clone 4C1 detects 3-beta-HSD I (HSD3B1) in human tissues. It targets an epitope with in 14 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.