Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD3B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576669 Supplier Invitrogen™ Supplier No. PA576669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

3 beta-hydroxysteroid dehydrogenase (3beta-HSD), also known as HSD3B1 or HSDB3, is a bifunctional enzyme that plays a crucial role in the synthesis of all classes of hormonal steroids. Two human 3beta-HSD proteins, designated type I (3beta-HSD) and type II (3beta-HSD2), are expressed by different genes and function in different areas of the body. Localized to the membrane of the endoplasmic reticulum (ER) and expressed in skin and placenta, 3beta-HSD is the type I protein that catalyzes the oxidative conversion of delta5-ene-3-beta-hydroxy steroid, as well as the conversion of various ketosteroids. Defects in the gene encoding 3beta-HSD are associated with classic salt wasting, genital ambiguity, hypogonadism, insulin-resistant polycystic ovary syndrome (PCOS) and an increased susceptibility to prostate cancer. Additionally, congenital deficiency of 3beta-HSD activity results in a severe depletion of steroid formation which can be lethal in young children.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD3B2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HSD3B2
Gene Accession No. P26149, P26439
Gene Alias 3 beta HSD II; 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase 2; 3 beta-hydroxysteroid dehydrogenase 6; 3 beta-hydroxysteroid dehydrogenase type II, delta 5-delta 4-isomerase type II, 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 4; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type IV; 3beta-HSD; 3-beta-HSD adrenal and gonadal type; 3-beta-HSD II; 3-beta-HSD IV; 3-beta-hydroxy-5-ene steroid dehydrogenase; 3-beta-hydroxy-Delta(5)-steroid dehydrogenase; delta 5-delta 4-isomerase type II; Delta-5-3-ketosteroid isomerase; HSD3B; Hsd3b1; Hsd3b2; Hsd3b6; HSD-3beta; HSDB; HSDB3B; Hydroxy-delta-5-steroid dehydrogenase 3 beta- and steroid delta-isomerase; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 6; hydroxysteroid dehydrogenase-2, delta>5>-3-beta; hydroxysteroid dehydrogenase-6 delta>5>-3-beta; progesterone reductase; SDR11E2; short chain dehydrogenase/reductase family 11E, member 2; Steroid Delta-isomerase
Gene Symbols HSD3B2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HSD3B1+ HSD3B2: 101-200.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15493, 3284
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.