Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSD3B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527775
Description
Recommended positive controls: HeLaS3, Molt-4. Predicted reactivity: Dog (80%), Cat (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
3 beta-hydroxysteroid dehydrogenase (3beta-HSD), also known as HSD3B1 or HSDB3, is a bifunctional enzyme that plays a crucial role in the synthesis of all classes of hormonal steroids. Two human 3beta-HSD proteins, designated type I (3beta-HSD) and type II (3beta-HSD2), are expressed by different genes and function in different areas of the body. Localized to the membrane of the endoplasmic reticulum (ER) and expressed in skin and placenta, 3beta-HSD is the type I protein that catalyzes the oxidative conversion of delta5-ene-3-beta-hydroxy steroid, as well as the conversion of various ketosteroids. Defects in the gene encoding 3beta-HSD are associated with classic salt wasting, genital ambiguity, hypogonadism, insulin-resistant polycystic ovary syndrome (PCOS) and an increased susceptibility to prostate cancer. Additionally, congenital deficiency of 3beta-HSD activity results in a severe depletion of steroid formation which can be lethal in young children.
Specifications
HSD3B2 | |
Polyclonal | |
Unconjugated | |
HSD3B2 | |
3 beta HSD II; 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase 2; 3 beta-hydroxysteroid dehydrogenase 6; 3 beta-hydroxysteroid dehydrogenase type II, delta 5-delta 4-isomerase type II, 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 4; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type IV; 3beta-HSD; 3-beta-HSD adrenal and gonadal type; 3-beta-HSD II; 3-beta-HSD IV; 3-beta-hydroxy-5-ene steroid dehydrogenase; 3-beta-hydroxy-Delta(5)-steroid dehydrogenase; delta 5-delta 4-isomerase type II; Delta-5-3-ketosteroid isomerase; HSD3B; Hsd3b1; Hsd3b2; Hsd3b6; HSD-3beta; HSDB; HSDB3B; Hydroxy-delta-5-steroid dehydrogenase 3 beta- and steroid delta-isomerase; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 6; hydroxysteroid dehydrogenase-2, delta>5>-3-beta; hydroxysteroid dehydrogenase-6 delta>5>-3-beta; progesterone reductase; SDR11E2; short chain dehydrogenase/reductase family 11E, member 2; Steroid Delta-isomerase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15493, 3284 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P26149, P26439 | |
HSD3B2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 216 of Human HSD3B2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction