Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD3B7 Polyclonal Antibody
HSD3B7 Polyclonal Antibody
Supplier: Thermo Scientific PA566350
Description
HSD3B7 Polyclonal Antibody for ICC/IF

Specifications
HSD3B7 | |
Polyclonal | |
Unconjugated | |
HSD3B7 | |
3 beta-hydroxy-delta 5-C27-steroid oxidoreductase; 3 beta-hydroxysteroid dehydrogenase type 7; 3 beta-hydroxysteroid dehydrogenase type VII; 3-beta-HSD VII; 3-beta-hydroxy-Delta(5)-C27 steroid oxidoreductase; AI195443; BB098564; C(27) 3-beta-HSD; C(27)-3BETA-HSD; CBAS1; Cca2; cholest-5-ene-3-be; cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase; Confluent 3Y1 cell-associated 2; HSD3B7; hsd3b7 {ECO:0000312; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7; PFIC4; RGD:628727}; SDR11E3; short chain dehydrogenase/reductase family 11E, member 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80270 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2F3 | |
HSD3B7 | |
Recombinant Human HSD3B7. Recombinant protein control fragment (Product #RP-107033). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction