Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSDL2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297749AF647
Description
HSDL2 Polyclonal specifically detects HSDL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HSDL2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
chromosome 9 open reading frame 99, EC 1.1.1, EC 1.1.1.100, FLJ25855, hydroxysteroid dehydrogenase like 2, member 1, MGC10940 | |
Produced in rabbits immunized with E. coli-derived Human HSDL2 fragment. | |
0.1 mL | |
Lipid and Metabolism | |
84263 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction