Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSDL2 Mouse anti-Human, Clone: 2A11BG6AH6, Abnova™

Catalog No. 89035493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-493 Supplier Abnova Corporation Supplier No. MAB14089

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human HSDL2.

  • High specificity
  • High reactivity

Specifications

Antigen HSDL2
Applications ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2A11BG6AH6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human HSDL2.
Dilution ELISA Immunocytochemistry Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. Q6YN16
Gene Alias C9orf99/FLJ25855/MGC10940/SDR13C1
Gene Symbols HSDL2
Host Species Mouse
Immunogen Human HSDL2.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84263
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.