Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSDL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100564 Supplier Invitrogen™ Supplier No. PA5100564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HSDL2.

HSDL2 is a protein coding gene. Has apparently no steroid dehydrogenase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSDL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSDL2
Gene Accession No. Q2TPA8, Q4V8F9, Q6YN16
Gene Alias 2610207I16Rik; C9orf99; Hsdl2; hydroxysteroid dehydrogenase like 2; hydroxysteroid dehydrogenase-like protein 2; RGD1305387; SDR13C1; short chain dehydrogenase/reductase family 13C member 1; short chain dehydrogenase/reductase family 13C, member 1
Gene Symbols HSDL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSDL2(Accession Q6YN16), corresponding to amino acid residues E360-L410.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313200, 72479, 84263
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.