Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSDL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598540
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598540 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598540 Supplier Invitrogen™ Supplier No. PA598540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSDL2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

HSDL2 is a protein coding gene. Has apparently no steroid dehydrogenase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSDL2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HSDL2
Gene Accession No. Q6YN16
Gene Alias 2610207I16Rik; C9orf99; Hsdl2; hydroxysteroid dehydrogenase like 2; hydroxysteroid dehydrogenase-like protein 2; RGD1305387; SDR13C1; short chain dehydrogenase/reductase family 13C member 1; short chain dehydrogenase/reductase family 13C, member 1
Gene Symbols HSDL2
Host Species Rabbit
Immunogen Recombinant Human Hydroxysteroid dehydrogenase-like protein 2 protein (1-345AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84263
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.