Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hSET1 Recombinant Rabbit Monoclonal Antibody (BLR024E), Bethyl Laboratories

Catalog No. 501571302
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1302 20 μL
50-157-1301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1302 Supplier Bethyl Laboratories, Inc Supplier No. A700024T
Add to Cart
Add to Cart

Rabbit Recombit Monoclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

SET1A is a component of a histone methyltransferase (HMT) complex that produces mono-, di-, and trimethylated histone H3 at Lys4. The complex is the analog of the S. cerevisiae Set1/COMPASS complex (Lee and Skalnik, 2005 [PubMed 16253997]).

Specifications

Antigen hSET1
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BLR024E
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene SETD1A
Gene Accession No. O15047
Gene Alias SET domain containing 1A, KMT2F, Set1A, Set1
Gene Symbols SETD1A
Host Species Rabbit
Immunogen Between 1200 and 1250
Purification Method Ion-exchange chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233904, 9739
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.