Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hSET1B Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572456
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2456 10 μL
50-157-2455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2456 Supplier Bethyl Laboratories, Inc Supplier No. IHC00641T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Rabbit, Gorilla, Chimpanzee, Northern white-cheeked gibbon and hirteen-lined ground squir

SETD1B is a component of the SET1 histone methyltransferase (HMT) complex. This complex specifically methylates lysine 4 of histone H3, but only if the neighboring lysine 9 residue is not yet methylated. Methylation of H3K4 represents a specific tag for epigenetic transcriptional activation. SETD1B shows a non-overlapping localization with SETD1A, suggesting that both proteins play a specific role in the epigenetic control of chromatin structure and gene expression.

Specifications

Antigen hSET1B
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SETD1B
Gene Accession No. Q9UPS6
Gene Alias SET1B, KMT2G, Histone-lysine N-methyltransferase SETD1B, KIAA1076, Lysine N-methyltransferase 2G, SET domain-containing protein 1B, hSET1B
Gene Symbols SETD1B
Host Species Rabbit
Immunogen Between 350 and 400
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23067
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.