Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1 Antibody (10H4), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP242205AF405
Description
HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
HSF1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HSF1 | |
Purified recombinant mouse HSF1 protein | |
0.1 mL | |
Primary | |
Detects approx 85kDa (unstressed cell lysates) and approx 95kDa (heat shocked cell lysates). | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
10H4 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
Rat | |
Protein G purified | |
Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
3297 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction