Learn More
HSF1 Antibody (4B4), DyLight 680, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP242206FR
Catalog No. NBP242206FR
Only null left
Add to Cart
Description
Description
HSF1 Monoclonal specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
HSF1
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
HSF1
Purified recombinant mouse HSF1 protein, epitope mapping to amino acids 425-439
0.1 mL
Primary
Detects approx 85kDa (unstressed cell lysates) and approx 95kDa (heat shocked cell lysates).
Store at 4C in the dark.
IgG1
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
4B4
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1
Rat
Protein G purified
Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
3297
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.