Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
Supplier: Novus Biologicals NBP1974750.05MG
Description
HSF1 Polyclonal specifically detects HSF1 in Human, Mouse, Rat, Gerbil, Primate, Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Gel Super Shift Assays, Enzyme Immunoassay.Specifications
| HSF1 | |
| Polyclonal | |
| Western Blot 1:1000 (colorimetric), ELISA, Immunoprecipitation 1:10-1:500, Gel Super Shift Assays 1:1-1:100, Enzyme Immunoassay | |
| Q00613 | |
| HSF1 | |
| Recombinant human HSF1. | |
| 0.05 mg | |
| Primary | |
| Recognizes nonphosphorylated HSF1 at ∼60kDa and phosphorylated HSF1 at ∼80kDa. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation, Gel Shift | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3297 | |
| Human, Mouse, Rat, Gerbil, Primate, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction