Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1, p Ser326 Antibody (SR2246), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HSF1 |
---|---|
Clone | SR2246 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunoprecipitationSpecifications
HSF1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
A synthesized peptide derived from human phospho-HSF1 [Ser326] (Uniprot #: Q00613 ) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR2246 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3297 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title