Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1 Rabbit anti-Human, Clone: S07-0G0, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314895100UL
This item is not returnable.
View return policy
Description
HSF1 Monoclonal antibody specifically detects HSF1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Supershift AssaySpecifications
HSF1 | |
Monoclonal | |
Unconjugated | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Shift | |
S07-0G0 | |
Western Blot, ELISA 1:100 - 1:2000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Supershift Assay | |
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
A synthetic peptide of human HSF1 (Uniprot#: Q00613) | |
100 μg | |
Cancer, DNA Repair, Transcription Factors and Regulators | |
3297 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction